Learn More
Quidel Amplivue™ C. difficile Assay Kit Accessories: Amplivue rack
SDP
Catalog No. 23043154 Shop All QuidelOrtho Products
Catalog No. 23-043-154 Supplier Quidel Supplier No. M208
Add to Cart
Add to Cart
Accessories for use with C. difficile assay kit used in detection of toxigenic C. difficile in stool samples.
  • 64°C amplification block with dry bath and metal module
  • Rack
Specifications
For Use With (Application) AmpliVue C. difficile Assay Kit Accessories
Product Type Amplivue rack