Learn More
Cayman Chemical AMPMaxSpec KIt 15test
SDP
Catalog No. 502896642
Encompass_Preferred
Cayman’s AMP+ MaxSpec Kit contains a positively charged reagent (AMP+) that can be coupled quantitatively to free carboxylic acids through an amide bond linkage. The resulting derivatized product thus contains a positively charged group that shows improved electrospray ionization and consequently improved MS sensitivity. The derivatization reagent is stable to MS/MS fragmentation and thus isobaric derivatives undergo compound-specific fragmentation to give unique fragment ions.
Catalog No. 50-289-6642 Supplier Cayman Chemical Supplier No. 71000015 TEST
Cayman’s AMP+ MaxSpec Kit contains a positively charged reagent (AMP+) that can be coupled quantitatively to free carboxylic acids through an amide bond linkage. The resulting derivatized product thus contains a positively charged group that shows improved electrospray ionization and consequently improved MS sensitivity. The derivatization reagent is stable to MS/MS fragmentation and thus isobaric derivatives undergo compound-specific fragmentation to give unique fragment ions.