Learn More
Cayman Chemical AMP+ MaxSpec Kit, 50 test
SDP
Catalog No. 502896644
Encompass_Preferred
Cayman’s AMP+ MaxSpec Kit contains a positively charged reagent (AMP+) that can be coupled quantitatively to free carboxylic acids through an amide bond linkage. The resulting derivatized product thus contains a positively charged group that shows improved electrospray ionization and consequently improved MS sensitivity. The derivatization reagent is stable to MS/MS fragmentation and thus isobaric derivatives undergo compound-specific fragmentation to give unique fragment ions.
Catalog No. 50-289-6644 Supplier Cayman Chemical Supplier No. 71000050 TEST
Cayman’s AMP+ MaxSpec Kit contains a positively charged reagent (AMP+) that can be coupled quantitatively to free carboxylic acids through an amide bond linkage. The resulting derivatized product thus contains a positively charged group that shows improved electrospray ionization and consequently improved MS sensitivity. The derivatization reagent is stable to MS/MS fragmentation and thus isobaric derivatives undergo compound-specific fragmentation to give unique fragment ions.