Learn More
Electron Microscopy Sciences Ampoule Breaker; Soft Silicone, Reusable
SDP
Catalog No. 50997515
Encompass_Preferred
Reusable Made from soft silicone. It offers a simple way to break pre-scored glass ampoules safely. Accommodates 5, 7, and 10 ml size ampoules. Reduces injury and spillage.
Catalog No. 50-997-515 Supplier Electron Microscopy Sciences Supplier No. 60600
Reusable Made from soft silicone. It offers a simple way to break pre-scored glass ampoules safely. Accommodates 5, 7, and 10 ml size ampoules. Reduces injury and spillage.