Learn More
Electron Microscopy Sciences Ampoule Cracker for 5-10 ml
SDP
Catalog No. 50997517
Encompass_Preferred
The Ampoule Cracker is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules. Simply slip the Ampoule Cracker over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Use: 5-10 ml
Catalog No. 50-997-517 Supplier Electron Microscopy Sciences Supplier No. 6060510
The Ampoule Cracker is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules. Simply slip the Ampoule Cracker over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Use: 5-10 ml