Learn More
Cayman Chemical AMPR22trIfluoroACTAte sal 10mg
SDP
Catalog No. 502919351
Encompass_Preferred
An antimicrobial peptide; active against S. aureus, MRSA, P. aeruginosa, and carbapenem-resistant P. aeruginosa (MBCs = 2, 2, 2, and 4 UG/ml, respectively); induces bacterial outer membrane permeability and ATP leakage to the extracellular space in S. aureus and P. aeruginosa; increases survival in a mouse model of sepsis induced by S. aureus at 10 mg/kg
Catalog No. 50-291-9351 Supplier Cayman Chemical Supplier No. 3695410 MG
An antimicrobial peptide; active against S. aureus, MRSA, P. aeruginosa, and carbapenem-resistant P. aeruginosa (MBCs = 2, 2, 2, and 4 UG/ml, respectively); induces bacterial outer membrane permeability and ATP leakage to the extracellular space in S. aureus and P. aeruginosa; increases survival in a mouse model of sepsis induced by S. aureus at 10 mg/kg