Learn More
Medchemexpress LLC Ampreloxetine hydrochloride | 1227056-87-2 | 99.5% | 357.80 | C18H19ClF3NO | 10 MG
SDP
Catalog No. 5000239902
Encompass_Preferred
Ampreloxetine hydrochloride is the hydrochloride salt of ampreloxetine (TD-9855), an orally active, CNS-penetrant inhibitor of norepinephrine and serotonin reuptake. Supplied as a research-grade small-molecule reagent, it is intended for preclinical pharmacology and transporter studies investigating NET and SERT function.
  • High reported purity (99.5%) suitable for research assays.
  • Provided as a solid for accurate weighing and formulation.
  • Suitable for in vitro and in vivo preclinical studies of monoamine transporters.
  • Well-characterized molecular properties (C18H19ClF3NO, MW 357.80).
  • Available in small milligram package sizes for dose-ranging and screening.
Catalog No. 50-002-39902 Supplier Medchemexpress LLC Supplier No. HY10712810MG
Ampreloxetine hydrochloride is the hydrochloride salt of ampreloxetine (TD-9855), an orally active, CNS-penetrant inhibitor of norepinephrine and serotonin reuptake. Supplied as a research-grade small-molecule reagent, it is intended for preclinical pharmacology and transporter studies investigating NET and SERT function.
  • High reported purity (99.5%) suitable for research assays.
  • Provided as a solid for accurate weighing and formulation.
  • Suitable for in vitro and in vivo preclinical studies of monoamine transporters.
  • Well-characterized molecular properties (C18H19ClF3NO, MW 357.80).
  • Available in small milligram package sizes for dose-ranging and screening.