Learn More
Medchemexpress LLC Ampreloxetine hydrochloride | 1227056-87-2 | 99.5% | 357.80 g/mol | C18H19ClF3NO | 100 MG
SDP
Catalog No. 5000240376
Encompass_Preferred
Ampreloxetine hydrochloride is an orally active, CNS-penetrant inhibitor of norepinephrine and serotonin transporters used as a research compound to investigate monoamine reuptake and central nervous system pharmacology. Supplied as the hydrochloride salt, it is intended for preclinical and biochemical assays and is characterized by high reported purity.
  • Orally active, CNS-penetrant norepinephrine and serotonin transporter inhibitor.
  • Useful for studies of monoamine reuptake and central nervous system pharmacology.
  • Provided as a hydrochloride salt to aid solubility in formulations.
  • High reported purity suitable for biochemical and preclinical assays.
  • Available in laboratory-scale quantity for research use.
  • Molecular weight 357.80 g/mol and formula C18H19ClF3NO.
Catalog No. 50-002-40376 Supplier Medchemexpress LLC Supplier No. HY107128100MG
Ampreloxetine hydrochloride is an orally active, CNS-penetrant inhibitor of norepinephrine and serotonin transporters used as a research compound to investigate monoamine reuptake and central nervous system pharmacology. Supplied as the hydrochloride salt, it is intended for preclinical and biochemical assays and is characterized by high reported purity.
  • Orally active, CNS-penetrant norepinephrine and serotonin transporter inhibitor.
  • Useful for studies of monoamine reuptake and central nervous system pharmacology.
  • Provided as a hydrochloride salt to aid solubility in formulations.
  • High reported purity suitable for biochemical and preclinical assays.
  • Available in laboratory-scale quantity for research use.
  • Molecular weight 357.80 g/mol and formula C18H19ClF3NO.