Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampreloxetine TFA | 1227056-85-0 | 98.3% | 25 MG
SDP

Catalog No. 5000234616
Encompass_Preferred

Ampreloxetine TFA (trifluoroacetate) is the TFA salt of ampreloxetine (TD-9855), an orally active inhibitor of norepinephrine and serotonin uptake supplied as a solid research reagent. It is intended for in vitro and preclinical research and is provided in small package sizes with recommended refrigerated storage.

  • Potent norepinephrine and serotonin uptake inhibitor.
  • Trifluoroacetate (TFA) salt, solid form.
  • Purity 98.3%.
  • Molecular weight 435.36 g/mol.
  • Available in small research sizes, including 25 mg.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-34616 Supplier Medchemexpress LLC Supplier No. HY119541A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ampreloxetine TFA (trifluoroacetate) is the TFA salt of ampreloxetine (TD-9855), an orally active inhibitor of norepinephrine and serotonin uptake supplied as a solid research reagent. It is intended for in vitro and preclinical research and is provided in small package sizes with recommended refrigerated storage.

  • Potent norepinephrine and serotonin uptake inhibitor.
  • Trifluoroacetate (TFA) salt, solid form.
  • Purity 98.3%.
  • Molecular weight 435.36 g/mol.
  • Available in small research sizes, including 25 mg.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.