Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampreloxetine trifluoroacetate | 1227056-85-0 | 98.3% | 5 MG
SDP

Catalog No. 5000233133
Encompass_Preferred

Ampreloxetine trifluoroacetate (TFA) is the trifluoroacetate salt of ampreloxetine, an orally active norepinephrine and serotonin (5-HT) uptake inhibitor investigated for research into neurogenic orthostatic hypotension. Supplied as a white to off-white solid with high purity for laboratory research use only.

  • High purity (98.3%) suitable for research applications.
  • Trifluoroacetate salt form to improve stability and solubility properties.
  • Molecular weight 435.36 g/mol; formula C20H19F6NO3.
  • Available in small research pack sizes (e.g., 5 MG) for assay development and preclinical studies.
  • Intended for laboratory research use only; not for human or animal therapeutic use.

Catalog No. 50-002-33133 Supplier Medchemexpress LLC Supplier No. HY119541A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ampreloxetine trifluoroacetate (TFA) is the trifluoroacetate salt of ampreloxetine, an orally active norepinephrine and serotonin (5-HT) uptake inhibitor investigated for research into neurogenic orthostatic hypotension. Supplied as a white to off-white solid with high purity for laboratory research use only.

  • High purity (98.3%) suitable for research applications.
  • Trifluoroacetate salt form to improve stability and solubility properties.
  • Molecular weight 435.36 g/mol; formula C20H19F6NO3.
  • Available in small research pack sizes (e.g., 5 MG) for assay development and preclinical studies.
  • Intended for laboratory research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.