Learn More
TARGETMOL CHEMICALS INC AMPRENAVIR 25MG
SDP
Catalog No. 502576524
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Amprenavir (KVX-478) is a synthetic derivative of hydroxyethylamine sulfonamide that selectively binds to and inhibits human immunodeficiency virus (HIV) protease. purity: 99%
Catalog No. 50-257-6524 Supplier TARGETMOL CHEMICALS INC Supplier No. T638825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Amprenavir (KVX-478) is a synthetic derivative of hydroxyethylamine sulfonamide that selectively binds to and inhibits human immunodeficiency virus (HIV) protease. purity: 99%