Learn More
Apexbio Technology LLC Amprenavir (agenerase), 50mg. Cas: 161814-49-9 MFCD: MFCD00934214. HIV-1 protease inhibitor
SDP
Catalog No. 501905075
Encompass_Preferred
Amprenavir (agenerase), an N,N-disubstituted hydroxyethylamino sulfonamide, is a novel inhibitor of human immunodeficiency virus type 1 (HIV-1) protease that potently inhibits the activity of HIV-1 protease with a value of 50% inhibition concentration IC50 of 0.6 nM Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5075 Supplier Apexbio Technology LLC Supplier No. A820150
Add to Cart
Add to Cart
Amprenavir (agenerase), an N,N-disubstituted hydroxyethylamino sulfonamide, is a novel inhibitor of human immunodeficiency virus type 1 (HIV-1) protease that potently inhibits the activity of HIV-1 protease with a value of 50% inhibition concentration IC50 of 0.6 nM Other sizes are available. Please inqury us for quote.