Learn More
Cayman Chemical Amprenavir-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2738376-78-6, 1 mg
SDP
Catalog No. 502938654
Encompass_Preferred
An internal standard for the quantification of amprenavir by GC- or LC-MS
Catalog No. 50-293-8654 Supplier Cayman Chemical Supplier No. 287031 MG
An internal standard for the quantification of amprenavir by GC- or LC-MS