Learn More
Cayman Chemical AmprenavIr-d4 500ug
SDP
Catalog No. 502938972
Encompass_Preferred
An internal standard for the quantification of amprenavir by GC- or LC-MS
Catalog No. 50-293-8972 Supplier Cayman Chemical Supplier No. 28703500 UG
An internal standard for the quantification of amprenavir by GC- or LC-MS