Learn More
Apexbio Technology LLC Amprolium HCl, 100mg Cas# 137-88-2
SDP
Supplier:  Apexbio Technology LLC B1663100
Encompass_Preferred
Amprolium HCl 50mg. 1431368-48-7. MFCD28124398 Amprolium chloride is a thiamin antagonist, which prevents carbohydrate synthesis by blocking thiamine uptake. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8632