Learn More
Medchemexpress LLC Amprolium hydrochloride | 137-88-2 | 98.79% | 250 MG
SDP
Catalog No. 5000407370
Encompass_Preferred
Amprolium hydrochloride is an anti-parasitic drug that is taken orally and acts as an antagonist to thiamine in intestinal absorption. It is for research use only and not sold to patients.
  • Acts as an antagonist to thiamine in intestinal absorption
  • Targets Coccidia
  • Suitable for in vivo and in vitro applications
  • For research use only
Catalog No. 50-004-07370 Supplier Medchemexpress LLC Supplier No. 250MGHYB0937A
Amprolium hydrochloride is an anti-parasitic drug that is taken orally and acts as an antagonist to thiamine in intestinal absorption. It is for research use only and not sold to patients.
  • Acts as an antagonist to thiamine in intestinal absorption
  • Targets Coccidia
  • Suitable for in vivo and in vitro applications
  • For research use only