Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amprolium (hydrochloride) | 137-88-2 | 98.8% | 1 G
SDP

Catalog No. 5000408372
Encompass_Preferred

Amprolium hydrochloride is an anti-parasitic drug that is taken orally and acts as an antagonist to thiamine in intestinal absorption. With a molecular formula of C14H20Cl2N4 and a molecular weight of 315.24, this compound is supplied as a white to off-white solid with a high purity of 98.79%.

  • Anti-parasitic drug targeting coccidia
  • Acts as a thiamine antagonist
  • High purity of 98.79%
  • White to off-white solid appearance
  • Soluble in water (≥ 200 mg/mL)
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-08372 Supplier Medchemexpress LLC Supplier No. HYB0937A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amprolium hydrochloride is an anti-parasitic drug that is taken orally and acts as an antagonist to thiamine in intestinal absorption. With a molecular formula of C14H20Cl2N4 and a molecular weight of 315.24, this compound is supplied as a white to off-white solid with a high purity of 98.79%.

  • Anti-parasitic drug targeting coccidia
  • Acts as a thiamine antagonist
  • High purity of 98.79%
  • White to off-white solid appearance
  • Soluble in water (≥ 200 mg/mL)
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.