Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amprolium (hydrochloride) | 137-88-2 | 98.8% | 1 ML
SDP

Catalog No. 5000408371
Encompass_Preferred

Amprolium hydrochloride is provided as a 10 mM solution in 1 mL, with a purity of 98.8%. This product is intended for research use only and should be handled by qualified personnel in appropriately equipped facilities.

  • 10 mM solution in 1 mL
  • Purity: 98.8% (HPLC)
  • Recommended storage: 4°C for sealed solutions, away from moisture
  • For longer-term storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if shipment duration is less than 2 weeks
  • Intended for research applications

Catalog No. 50-004-08371 Supplier Medchemexpress LLC Supplier No. HYB0937A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amprolium hydrochloride is provided as a 10 mM solution in 1 mL, with a purity of 98.8%. This product is intended for research use only and should be handled by qualified personnel in appropriately equipped facilities.

  • 10 mM solution in 1 mL
  • Purity: 98.8% (HPLC)
  • Recommended storage: 4°C for sealed solutions, away from moisture
  • For longer-term storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if shipment duration is less than 2 weeks
  • Intended for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.