Learn More
Medchemexpress LLC Amprolium (hydrochloride) | 137-88-2 | 98.8% | 500 MG
SDP
Catalog No. 5000408289
Encompass_Preferred
Amprolium hydrochloride is an anti-parasitic drug administered orally, functioning as a thiamine antagonist in intestinal absorption. It has been used as a preventive measure, particularly when combined with other anti-coccidial drugs. Its antagonistic effect on thiamine absorption primarily occurs at the intestinal epithelial cells. This product is for research use only and has not been fully validated for medical applications.
  • Anti-parasitic drug
  • Administered orally
  • Acts as a thiamine antagonist in intestinal absorption
  • Used for preventive measures against coccidiosis
  • Targets coccidia
  • Antagonistic effect primarily at intestinal epithelial cells
  • For research use only
Catalog No. 50-004-08289 Supplier Medchemexpress LLC Supplier No. HYB0937A500MG
Amprolium hydrochloride is an anti-parasitic drug administered orally, functioning as a thiamine antagonist in intestinal absorption. It has been used as a preventive measure, particularly when combined with other anti-coccidial drugs. Its antagonistic effect on thiamine absorption primarily occurs at the intestinal epithelial cells. This product is for research use only and has not been fully validated for medical applications.
  • Anti-parasitic drug
  • Administered orally
  • Acts as a thiamine antagonist in intestinal absorption
  • Used for preventive measures against coccidiosis
  • Targets coccidia
  • Antagonistic effect primarily at intestinal epithelial cells
  • For research use only