Learn More
Research Products International Corp AMPSO, 25 G
SDP
Catalog No. 501978435
Encompass_Preferred
  • pKa (25°C): 9.0
  • The working pH range in aqueous solution is 8.3 - 9.7
Catalog No. 50-197-8435 Supplier Research Products International Corp Supplier No. A2052025.0
  • pKa (25°C): 9.0
  • The working pH range in aqueous solution is 8.3 - 9.7