Learn More
Research Products International Corp AMPSO [3-((1,1-Dimethyl-2-hydroxyethyl)amino)-2-hydroxypropanesulfonic acid], 100 Grams
SDP
Catalog No. 50488487
Encompass_Preferred
  • pKa (25°C): 9.0
  • The working pH range in aqueous solution is 8.3 - 9.7
Catalog No. 50-488-487 Supplier Research Products International Corp Supplier No. A20520100.0
  • pKa (25°C): 9.0
  • The working pH range in aqueous solution is 8.3 - 9.7