Learn More
Agilent Technologies AMPTABS AMPICILLIN TABLETS
Catalog No. 50125514
Encompass
AmpTabs Ampicillin Tablets are used to screen for ampicillin-resistant bacterial colonies on an ampicillin-medium.
Catalog No. 50-125-514 Supplier Agilent Technologies Supplier No. 300021
AmpTabs Ampicillin Tablets are used to screen for ampicillin-resistant bacterial colonies on an ampicillin-medium.