Learn More
Sigma Aldrich Fine Chemicals Biosciences Ampule breaker/collar for
Catalog No. 502899992
Encompass
How to Use1. Slip polyethylene breaker/ collar over top of ampule.2. Grip bottom of ampule with one hand and breaker/collar with other.3. Gently snap top of ampule off.4. Discard collar and ampule top.
Catalog No. 50-289-9992 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z122890100EA
How to Use1. Slip polyethylene breaker/ collar over top of ampule.2. Grip bottom of ampule with one hand and breaker/collar with other.3. Gently snap top of ampule off.4. Discard collar and ampule top.