Learn More
AdipoGen AMQI
SDP
Supplier:  AdipoGen CDXA0036M100
Encompass_Preferred
Chemical. CAS 7270-83-9. Formula C12H12INO2. MW 329.13. Synthetic. Fluorogenic substrate for cholinesterase. Non-fluorescent 7-acetoxy-1-methyl quinolinium iodide is hydrolysed to the highly fluorescent 7-hydroxy-1-methyl quinolinium iodide.
Catalog No. 50-256-1855