Learn More
Electron Microscopy Sciences SEM Sample Holder Amray Base Universal Holder With Pin C
SDP
Catalog No. 5028817
Encompass_Preferred
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. Holds any sample from 3mm to 29mm. Four removable arms. Easy sample insert and removal. Made from aluminum. Base measures: 48mm x42mm x12mm thick. Pin overall measures: 34.5mm x 16mm dia
Catalog No. 50-288-17 Supplier Electron Microscopy Sciences Supplier No. 7591003
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. Holds any sample from 3mm to 29mm. Four removable arms. Easy sample insert and removal. Made from aluminum. Base measures: 48mm x42mm x12mm thick. Pin overall measures: 34.5mm x 16mm dia