Learn More
Enterprise Technology Solutions Amrep,Inc. Dust Mop Treatment, Lemon/Citrus, 16oz, White
SDP
Catalog No. 5000790089
Encompass_Preferred
Aspire Dust Mop Treatment replaces sweeping compounds and eliminates the dust created when using mophead-equipped push brooms. The advanced, professional-strength formula contains no wax or silicone, does not make the treated surface slippery and will not damage floor surfaces. Use on asphalt tile, linoleum, vinyl, rubber tile, marble, granite, natural stone, ceramic, concrete, quarry tile, magnesite, cork and wood including polyurethane-coated wood such as gymnasium floors, racquet courts and more. It can also be used on flooring coated with removable polymer finish. Cloths treated with this product are ideal for dusting plastic, laminates, chalkboards and more. The nonflammable, water-based formula also features a pleasant light lemon fragrance. It is 98 percent VOC-free.
Catalog No. 50-007-90089 Supplier Enterprise Technology Solutions Supplier No. AMR1038049
Aspire Dust Mop Treatment replaces sweeping compounds and eliminates the dust created when using mophead-equipped push brooms. The advanced, professional-strength formula contains no wax or silicone, does not make the treated surface slippery and will not damage floor surfaces. Use on asphalt tile, linoleum, vinyl, rubber tile, marble, granite, natural stone, ceramic, concrete, quarry tile, magnesite, cork and wood including polyurethane-coated wood such as gymnasium floors, racquet courts and more. It can also be used on flooring coated with removable polymer finish. Cloths treated with this product are ideal for dusting plastic, laminates, chalkboards and more. The nonflammable, water-based formula also features a pleasant light lemon fragrance. It is 98 percent VOC-free.