Learn More
Enterprise Technology Solutions Amrep,Inc. Dust Treatment/Cleaner,RTU,f/Dust Mop,1 Gal,4/CT,CL
SDP
Catalog No. 5000790088
Encompass_Preferred
Apply Crystal Clear Deep Mop Treatment to dust mopheads to aid in the removal of dust, dirt and other abrasive matter from floor surfaces. Mopheads that are treated with this compound act as a "dust magnet" to attract and hold particles until mop is cleaned or laundered. It can also be used to treat cloths, mats and HVAC filters. This crystal-clear dressing with a grapefruit fragrance is non-oily and leaves a dry, protective, plasticized film on the surface of wood, asphalt, linoleum, vinyl, vinyl composition, rubber, terrazzo, concrete and gymnasium floors. Ready-to-use, low-odor treatment will not harm waxed floors.
Catalog No. 50-007-90088 Supplier Enterprise Technology Solutions Supplier No. AMR1003411
Apply Crystal Clear Deep Mop Treatment to dust mopheads to aid in the removal of dust, dirt and other abrasive matter from floor surfaces. Mopheads that are treated with this compound act as a "dust magnet" to attract and hold particles until mop is cleaned or laundered. It can also be used to treat cloths, mats and HVAC filters. This crystal-clear dressing with a grapefruit fragrance is non-oily and leaves a dry, protective, plasticized film on the surface of wood, asphalt, linoleum, vinyl, vinyl composition, rubber, terrazzo, concrete and gymnasium floors. Ready-to-use, low-odor treatment will not harm waxed floors.