Learn More
Enterprise Technology Solutions Amrep,Inc. Optimax Neutral Floor Cleaner, Lemon Sent, Green
SDP
Catalog No. 5000832698
Encompass_Preferred
High-performance floor cleaner is safe for resilient tile, stone, masonry and sealed wood. Neutral pH won't harm floor finishes. Advanced surfactant technology is excellent for breaking up and dispersing even stubborn grease, dirt, and oil deposits from flooring surfaces either by hand or through automatic scrubbing machines. Environmentally preferable formula is DfE approved. Alternative ingredients result in a more positive environmental and human health profile. Concentrated cleaner is economical to use with a high dilution rate of 1:128. The pleasant lemon scent makes it easy to work with and around.
Catalog No. 50-008-32698 Supplier Enterprise Technology Solutions Supplier No. AMR1033704
High-performance floor cleaner is safe for resilient tile, stone, masonry and sealed wood. Neutral pH won't harm floor finishes. Advanced surfactant technology is excellent for breaking up and dispersing even stubborn grease, dirt, and oil deposits from flooring surfaces either by hand or through automatic scrubbing machines. Environmentally preferable formula is DfE approved. Alternative ingredients result in a more positive environmental and human health profile. Concentrated cleaner is economical to use with a high dilution rate of 1:128. The pleasant lemon scent makes it easy to work with and around.