Learn More
TARGETMOL CHEMICALS INC AMRI-59 5MG
SDP
Catalog No. 502580684
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMRI-59 is a potent inhibitor of PrxI with anti-tumor activity. purity: 97%
Catalog No. 50-258-0684 Supplier TARGETMOL CHEMICALS INC Supplier No. T266235MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMRI-59 is a potent inhibitor of PrxI with anti-tumor activity. purity: 97%