Learn More
Medchemexpress LLC Amrubicin hydrochloride | 110311-30-3 | MFCD00876391 | 98.78% | C25H26ClNO9 | 1 MG
SDP
Catalog No. 5000432519
Encompass_Preferred
Amrubicin hydrochloride (SM-5887) is a DNA topoisomerase II inhibitor primarily used for cancer research. It exhibits significant cytotoxic effects on various cell lines, including A549, HT-29, and K562 cells, and shows potent antitumor activities in SCLC tumors and other cancer xenograft models.
  • DNA topoisomerase II inhibitor
  • Used for cancer research
  • Cytotoxic against A549, HT-29, and K562 cells
  • Shows radio-enhancement effects on human lung adenocarcinoma A549 cells
  • Antitumor activities against SCLC tumors
Catalog No. 50-004-32519 Supplier Medchemexpress LLC Supplier No. HYB0067A1MG
Amrubicin hydrochloride (SM-5887) is a DNA topoisomerase II inhibitor primarily used for cancer research. It exhibits significant cytotoxic effects on various cell lines, including A549, HT-29, and K562 cells, and shows potent antitumor activities in SCLC tumors and other cancer xenograft models.
  • DNA topoisomerase II inhibitor
  • Used for cancer research
  • Cytotoxic against A549, HT-29, and K562 cells
  • Shows radio-enhancement effects on human lung adenocarcinoma A549 cells
  • Antitumor activities against SCLC tumors