Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AMS-4 Beta Air Monitor

Catalog No. 19175228 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Display unit without sampling head
Integral vacuum pump w/115 V AC, 60 Hz
Radial entry sampling head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-175-228 Display unit without sampling head
19-175-229 Radial entry sampling head
19-175-230 Integral vacuum pump w/115 V AC, 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-175-228 Supplier Thermo Scientific™ Supplier No. AMS4

missing translation for 'validMainframeMsg'

Provide an early warning to workers exposed to potential airborne releases of beta-emitting particulates when using Thermo Scientific™ AMS-4 Beta Air Monitor.

Provide an early warning to workers exposed to potential airborne releases of beta-emitting particulates when using Thermo Scientific™ AMS-4 Beta Air Monitor. Lightweight and robust design accommodates both fixed and portable use applications.

Continuous air monitor (CAM) offering beta particulate, radioiodine, or noble gas detection.
  • Detachable sampling head
  • Reports both DAC and cpm
  • Real time gamma background subtraction
  • Stand alone or network configuration
  • Mass flow measurements
  • Fast and slow concentration alarms, cpm alarm, DAC-hour alarm
  • Computer calibration
  • Can also be used for stack monitoring of effluent releases using the in-line detector head
  • Keypad: 18 keys, full numeric plus function keys
  • Status Indicators: Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
  • Printer: RS-232 serial DB9
  • Data Log Buffer: 2000 entries of combined Concentration and Status
  • Change information: Equivalent to 83 days of 1 hour detail, nearly 14 days of 10 mintes detail or over 24 hours of 1 minute detail
  • Alarm/Fail Relays: SPST, 15A at 30VDC/120VAC resistive
  • Analog Output: 0 to 5VDC output, five decade logarithmic

Compatible with:

Thermo Scientific ViewPoint Enterprise

TRUSTED_SUSTAINABILITY

Specifications

Alarm/Fail Relays SPST, 15 A at 30 VDC/120 VAC resistive
Certifications/Compliance Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Compatibility Thermo Scientific ViewPoint Enterprise
Connections Computer: DB9, RS-232 standard, RS-485 optional
Data Log Buffer 2000 entries of combined Concentration and Status
Description Display unit without sampling head
Alpha/Beta/Gamma Measurement Beta Measurement
For Use With (Application) Beta Monitoring
Detector Type Sealed proportional window
Flow Rate 8.5 to 113 L/min.
Outputs 0 to 5 VDC, five decade logarithmic
Radiation Type Beta
Display Type Two rows x 20 characters high visibility vacuum fluorescent, percentage of Alarm 40-element LED bar graph
Product Type Beta Air Monitor Display Unit
Keypad 18 keys, full numeric plus function keys
Status Indicator Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
Window Density 2 to 3 mg/cm²
Width (English) 11 in.
Height (English) 8.75 in.
Height (Metric) 22.2 cm
Depth (English) 2.8 in.
Diameter (Metric) Detector Window 50.8 mm
Weight (English) 7.5 lb.
Weight (Metric) 3.4 kg
Electrical Requirements 100 to 240 V 50/60 Hz
Voltage 100 V to 240 V
Wattage 120 W
Frequency 50/60 Hz
Width (Metric) 27.9 cm
Depth (Metric) 32.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.