Learn More
Perkin Elmer US LLC AMS Gas Line for NexION 2000
Catalog No. 502112172
Encompass_Preferred
The AMS spray chamber gas line is included in the NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150035) and the Productivity Configuration's Sample Introduction Kit (N8150036).
Catalog No. 50-211-2172 Supplier Perkin Elmer US LLC Supplier No. N8152374
The AMS spray chamber gas line is included in the NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150035) and the Productivity Configuration's Sample Introduction Kit (N8150036).