Learn More
Electron Microscopy Sciences AMS Series Sorbothane Balance Platform, 30 x 45 cm, Stainless Steel Surface
SDP
Catalog No. 5034491
Encompass_Preferred
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7Hz. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements. With four 7Hz sorbothane isolators which give an excellent 95 percent isolation at 50Hz.
Size: 30 x 45 cm
Catalog No. 50-344-91 Supplier Electron Microscopy Sciences Supplier No. 6870
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7Hz. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements. With four 7Hz sorbothane isolators which give an excellent 95 percent isolation at 50Hz.
Size: 30 x 45 cm