Learn More
Selleck Chemical LLC Amsacrine E4925-1G
SDP
Catalog No. 5000041781
Encompass_Preferred
Amsacrine(m-AMSA acridinyl anisidide) is an inhibitor of topoisomerase II Amsacrine acts as an antineoplastic agent which intercalates into the DNA of tumor cells it is also used in the treatment of acute myelogenous leukemia
Catalog No. 50-000-41781 Supplier Selleck Chemical LLC Supplier No. E49251G
Amsacrine(m-AMSA acridinyl anisidide) is an inhibitor of topoisomerase II Amsacrine acts as an antineoplastic agent which intercalates into the DNA of tumor cells it is also used in the treatment of acute myelogenous leukemia