Learn More
Abcam Amsacrine, DNA topoisomerase II inhibitor, 50MG
Catalog No. 502706192
Encompass_Preferred
MW 393.5 Da, Purity >98%. DNA topoisomerase II inhibitor. Antineoplastic agent. Intercalating planar polycyclic aromatic molecule which is active against a number of tumours in mice. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6192 Supplier Abcam Supplier No. AB14274250MG
MW 393.5 Da, Purity >98%. DNA topoisomerase II inhibitor. Antineoplastic agent. Intercalating planar polycyclic aromatic molecule which is active against a number of tumours in mice. Active in vivo..
The product is subject to the following: Abcam Restricted Use Statement