Learn More
TARGETMOL CHEMICALS INC AMSACRINE HYDROCHLORIDE 200MG
SDP
Catalog No. 502576658
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Amsacrine hydrochloride (acridinyl anisidide hydrochloride) is topoisomerase II inhibitor is used in the treatment of acute myelogenous leukemia. purity: 99%
Catalog No. 50-257-6658 Supplier TARGETMOL CHEMICALS INC Supplier No. T5820200MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Amsacrine hydrochloride (acridinyl anisidide hydrochloride) is topoisomerase II inhibitor is used in the treatment of acute myelogenous leukemia. purity: 99%