Learn More
Selleck Chemical LLC Amsacrine hydrochloride 25mg 54301-15-4 m-AMSA, acridinyl anisidide
SDP
Supplier:  Selleck Chemical LLC S562725MG
Encompass_Preferred
Amsacrine hydrochloride (m-AMSA, acridinyl anisidide) is the hydrochloride salt form of amsacrine, which is an inhibitor of topoisomerase II with antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1774114