Learn More
Medchemexpress LLC HY-13551A 50mg Medchemexpress, Amsacrine (hydrochloride) CAS:54301-15-4 Purity:>98%
SDP
Catalog No. 502081390
Encompass_Preferred
Medchemexpress, HY-13551A 50mg Amsacrine (hydrochloride) CAS:54301-15-4 Amsacrine hydrochloride (m-AMSA hydrochloride; acridinyl anisidide hydrochloride) is an inhibitor of topoisomerase II, and acts as an antineoplastic agent which can intercalates into the DNA of tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1390 Supplier Medchemexpress LLC Supplier No. HY13551A50MG
Medchemexpress, HY-13551A 50mg Amsacrine (hydrochloride) CAS:54301-15-4 Amsacrine hydrochloride (m-AMSA hydrochloride; acridinyl anisidide hydrochloride) is an inhibitor of topoisomerase II, and acts as an antineoplastic agent which can intercalates into the DNA of tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.