Learn More
Medchemexpress LLC Amsacrine hydrochloride | 54301-15-4 | MFCD07799963 | 99.9% | 429.92 g/mol | C21H20ClN3O3S | 5 MG
SDP
Catalog No. 5000222007
Encompass_Preferred
Amsacrine hydrochloride is a topoisomerase II inhibitor supplied as a research-grade analytical standard. It intercalates DNA and stabilizes topoisomerase II-DNA cleavage complexes, making it suitable for biochemical assays and cell biology studies of DNA damage, cell cycle, and autophagy. Available as a solid and as a DMSO solution, the material is characterized for consistent analytical performance.
Catalog No. 50-002-22007 Supplier Medchemexpress LLC Supplier No. HY13551A5MG
Amsacrine hydrochloride is a topoisomerase II inhibitor supplied as a research-grade analytical standard. It intercalates DNA and stabilizes topoisomerase II-DNA cleavage complexes, making it suitable for biochemical assays and cell biology studies of DNA damage, cell cycle, and autophagy. Available as a solid and as a DMSO solution, the material is characterized for consistent analytical performance.