Learn More
Cayman Chemical Amsacrine (hydrochloride), Purity: ≥98%, CAS: 54301-15-4, 10 mg
SDP
Catalog No. 502964154
Encompass_Preferred
A topoisomerase II poison; stimulates formation of a complex between DNA and topoisomerase II in which the DNA is cleaved and topoisomerase II remains covalently linked to the 5’ end of the DNA cleavage products; inhibits the DNA strand-passing activity of topoisomerase II at a concentration of 20 μg/ml; reduces growth of LoVo human colorrectal carcinoma and T1 human lymphoma cells in a dose-dependent manner; antitumor activity against several murine leukemia, melanoma, lung, colon, and mammary xenograft mouse models
Catalog No. 50-296-4154 Supplier Cayman Chemical Supplier No. 2222310 MG
A topoisomerase II poison; stimulates formation of a complex between DNA and topoisomerase II in which the DNA is cleaved and topoisomerase II remains covalently linked to the 5’ end of the DNA cleavage products; inhibits the DNA strand-passing activity of topoisomerase II at a concentration of 20 μg/ml; reduces growth of LoVo human colorrectal carcinoma and T1 human lymphoma cells in a dose-dependent manner; antitumor activity against several murine leukemia, melanoma, lung, colon, and mammary xenograft mouse models