Learn More
MSC AmScope Stereo Microscope, 3.5X-45X Zoom, Binocular, 64-LED Light
Catalog No. 5000513351
Encompass
This stereo microscope offers a powerful 3.5X-45X zoom magnification, ideal for detailed inspection and dissecting tasks. Featuring a binocular eyepiece with 10x magnification, it provides an upright image for comfortable and extended viewing sessions. The integrated 64-LED light ensures bright and uniform illumination of the specimen.
  • Offers 3.5X-45X zoom magnification for detailed viewing.
  • Equipped with a binocular eyepiece for comfortable observation.
  • Features 10x eyepiece magnification.
  • Provides an upright image direction.
  • Includes a 64-LED light for bright illumination.
Catalog No. 50-005-13351 Supplier MSC Supplier No. 13608211
This stereo microscope offers a powerful 3.5X-45X zoom magnification, ideal for detailed inspection and dissecting tasks. Featuring a binocular eyepiece with 10x magnification, it provides an upright image for comfortable and extended viewing sessions. The integrated 64-LED light ensures bright and uniform illumination of the specimen.
  • Offers 3.5X-45X zoom magnification for detailed viewing.
  • Equipped with a binocular eyepiece for comfortable observation.
  • Features 10x eyepiece magnification.
  • Provides an upright image direction.
  • Includes a 64-LED light for bright illumination.