Learn More
Electron Microscopy Sciences Unique Sorbothane built in Pharma grade stainless Steel platform removes up to 98 percent of unwanted vibrations. The custom damped Sorbothane isolator with a load capacity of 75 kg is resistant to almost all forms of chemical attack.
SDP
Catalog No. NC2946824
Encompass_Preferred
Sorbothane elastomer isolators have outstanding damping performance compared to isolators that use springs, rubber, or neoprene. With a load capacity of 75 kg they eliminate any rocking or wobble. Sorbothane rapidly damps down disturbances to the microscope, such as when it is being touched during operation. It greatly improves image quality by protecting microscopes from both horizontal and vertical vibrations emanating from sources such as floors, walls and benching and caused by traffic, elevators, rotating machinery and numerous other sources. The seamless design and pharma grade stainless steel featured in the AMS, AMSM, and AMSR Series makes these platforms ideal for use in laboratories, cleanrooms and in Medical and Biological environments.
Catalog No. NC2946824 Supplier Electron Microscopy Sciences Supplier No. 687175
Sorbothane elastomer isolators have outstanding damping performance compared to isolators that use springs, rubber, or neoprene. With a load capacity of 75 kg they eliminate any rocking or wobble. Sorbothane rapidly damps down disturbances to the microscope, such as when it is being touched during operation. It greatly improves image quality by protecting microscopes from both horizontal and vertical vibrations emanating from sources such as floors, walls and benching and caused by traffic, elevators, rotating machinery and numerous other sources. The seamless design and pharma grade stainless steel featured in the AMS, AMSM, and AMSR Series makes these platforms ideal for use in laboratories, cleanrooms and in Medical and Biological environments.