Learn More
Medchemexpress LLC AMT-NHS | 2925268-86-4 | 98.7% | 443.47 g·mol⁻¹ | C22H21NO7S | 10 MG
SDP
Catalog No. 5000286667
Encompass_Preferred
AMT-NHS is an RNA-protein crosslinker that combines a psoralen intercalator with an N-hydroxysuccinimide (NHS) ester to covalently capture RNA-protein interactions. It is cell-permeable and has been used in living cells to map diverse RNA-protein contacts, producing distinct crosslinking patterns for single- and double-stranded RNA regions.
  • Enables dual reactivity: psoralen-mediated photo-crosslinking of RNA and NHS-ester conjugation to protein amines.
  • Cell-permeable for in vivo crosslinking applications.
  • Soluble in DMSO at high concentration; may require warming and sonication to dissolve.
  • High supplied purity supports reproducible crosslinking experiments.
  • Stable as a powder under low-temperature storage; solution storage times are defined for sample handling.
Catalog No. 50-002-86667 Supplier Medchemexpress LLC Supplier No. HY15001410MG
AMT-NHS is an RNA-protein crosslinker that combines a psoralen intercalator with an N-hydroxysuccinimide (NHS) ester to covalently capture RNA-protein interactions. It is cell-permeable and has been used in living cells to map diverse RNA-protein contacts, producing distinct crosslinking patterns for single- and double-stranded RNA regions.
  • Enables dual reactivity: psoralen-mediated photo-crosslinking of RNA and NHS-ester conjugation to protein amines.
  • Cell-permeable for in vivo crosslinking applications.
  • Soluble in DMSO at high concentration; may require warming and sonication to dissolve.
  • High supplied purity supports reproducible crosslinking experiments.
  • Stable as a powder under low-temperature storage; solution storage times are defined for sample handling.