Learn More
Electron Microscopy Sciences AMT Series Laboratory Side Work Table, 120 x 75 cm, White Epoxy Surface
SDP
Catalog No. 5034497
Encompass_Preferred
These laboratory tables feature a rigid frame with levelling feet which ensure the table is in excellent contact with the floor. Available with White Epoxy, Stainless Steel or Trespa top. Options include : monitor arms and castor feet.
Size: 120 x 75 cm
Surface: White Epoxy
Catalog No. 50-344-97 Supplier Electron Microscopy Sciences Supplier No. 6920
These laboratory tables feature a rigid frame with levelling feet which ensure the table is in excellent contact with the floor. Available with White Epoxy, Stainless Steel or Trespa top. Options include : monitor arms and castor feet.
Size: 120 x 75 cm
Surface: White Epoxy