Learn More
Apexbio Technology LLC AMTB hydrochloride 1mg
SDP
Supplier:  Apexbio Technology LLC B75591
Encompass_Preferred
AMTB hydrochloride (CAS 926023-82-7) is a selective antagonist of the TRPM8 ion channel effectively inhibiting TRPM8-mediated activation induced by agonists such as icilin with a reported pIC50 of approximately 6 23 In addition to its primary action AMTB hydrochloride also exhibits non-selective inhibitory effects on voltage-gated sodium channels (NaV) This compound is commonly utilized in biomedical research to investigate the role of TRPM8 channel activity alterations in the pathophysiology of bladder overactivity and bladder pain syndrome
Catalog No. 50-000-88886