Learn More
Medchemexpress LLC Amtb hydrochloride | 926023-82-7 | >98.0% | 430.99 | C23H27ClN2O2S | 1 ML
SDP
Catalog No. 5000214100
Encompass_Preferred
AMTB hydrochloride is a small-molecule TRPM8 channel blocker used in laboratory pharmacology and ion channel research. It has been applied in studies of overactive bladder and painful bladder syndrome and is reported to affect voltage-gated sodium channels in some assays.
  • Selective TRPM8 channel blocker for ion channel studies.
  • Reported activity in research on overactive bladder and painful bladder syndrome.
  • Also inhibits voltage-gated sodium channels in some assays.
  • Soluble in DMSO; manufacturer provides stock solution preparation for concentrations such as 10 mM.
  • High purity available (≥98% by HPLC) and characterized by molecular weight 430.99.
Catalog No. 50-002-14100 Supplier Medchemexpress LLC Supplier No. HY10034510MM 1ML
AMTB hydrochloride is a small-molecule TRPM8 channel blocker used in laboratory pharmacology and ion channel research. It has been applied in studies of overactive bladder and painful bladder syndrome and is reported to affect voltage-gated sodium channels in some assays.
  • Selective TRPM8 channel blocker for ion channel studies.
  • Reported activity in research on overactive bladder and painful bladder syndrome.
  • Also inhibits voltage-gated sodium channels in some assays.
  • Soluble in DMSO; manufacturer provides stock solution preparation for concentrations such as 10 mM.
  • High purity available (≥98% by HPLC) and characterized by molecular weight 430.99.