Learn More
Medchemexpress LLC AMTB hydrochloride | 926023-82-7 | 99.2% | 430.99 | C23H27ClN2O2S | 5 MG
SDP
Catalog No. 5000214500
Encompass_Preferred
AMTB hydrochloride is a research-grade small molecule that selectively blocks TRPM8 ion channels and also inhibits voltage-gated sodium channels. Supplied as a white to off-white solid with high chemical purity, it is intended for in-vitro and in-vivo research applications investigating TRPM8-mediated physiology and pharmacology.
  • Selective TRPM8 channel blocker and non-selective voltage-gated sodium channel inhibitor.
  • High purity: 99.18%.
  • White to off-white solid form suitable for formulation.
  • Soluble in DMSO (up to 100 mg/mL) with sonication.
  • In vivo formulation protocols available for concentrations ≥ 2.5 mg/mL.
  • Supplied in small research pack sizes, including 5 mg.
Catalog No. 50-002-14500 Supplier Medchemexpress LLC Supplier No. HY1003455MG
AMTB hydrochloride is a research-grade small molecule that selectively blocks TRPM8 ion channels and also inhibits voltage-gated sodium channels. Supplied as a white to off-white solid with high chemical purity, it is intended for in-vitro and in-vivo research applications investigating TRPM8-mediated physiology and pharmacology.
  • Selective TRPM8 channel blocker and non-selective voltage-gated sodium channel inhibitor.
  • High purity: 99.18%.
  • White to off-white solid form suitable for formulation.
  • Soluble in DMSO (up to 100 mg/mL) with sonication.
  • In vivo formulation protocols available for concentrations ≥ 2.5 mg/mL.
  • Supplied in small research pack sizes, including 5 mg.