Learn More
TARGETMOL CHEMICALS INC Amthamine 2HCl 1MG
SDP
Catalog No. 502876759
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.5-(2-aminoethyl)-4-methyl-1,3-thiazol-2-amine dihydrochloride is a potent and selective antagonist of metabotropic glutamate receptor 5 (mGluR5). It has been shown to be effective in several neurological disorders, including fragile X syndrome, autism spectrum disorders, and addiction. Purity 95%
Catalog No. 50-287-6759 Supplier TARGETMOL CHEMICALS INC Supplier No. T501121MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.5-(2-aminoethyl)-4-methyl-1,3-thiazol-2-amine dihydrochloride is a potent and selective antagonist of metabotropic glutamate receptor 5 (mGluR5). It has been shown to be effective in several neurological disorders, including fragile X syndrome, autism spectrum disorders, and addiction. Purity 95%