Learn More
Medchemexpress LLC Amthamine dihydrobromide | 142457-00-9 | MFCD00673891 | 99.6% | 319.07 g/mol | C6H13Br2N3S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10106310MM 1ML
Encompass_Preferred
Amthamine dihydrobromide is a histamine receptor agonist supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material for laboratory use. It is used in receptor pharmacology and hepatotoxicity research to probe histamine-mediated biological responses.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.6%) suitable for biological assays.
  • Also available as solid in multiple vial sizes for synthesis or formulation.
  • CAS number 142457-00-9 for unambiguous identification.
  • Useful for histamine receptor pharmacology and hepatotoxicity studies.
Catalog No. 50-002-12272