Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amthamine dihydrobromide | 142457-00-9 | MFCD00673891 | 99.6% | 319.07 g/mol | C6H13Br2N3S | 10 MG
SDP

Catalog No. 5000212062
Encompass_Preferred

Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a histamine receptor agonist active at H1-H4 subtypes. It is supplied for research use to investigate histamine receptor pharmacology, gastric acid secretion, and hepatotoxicity models. Follow standard laboratory safety and storage recommendations.

  • High purity (99.6%) suitable for biochemical and pharmacological studies.
  • Molecular weight 319.07 g/mol.
  • Chemical formula C6H13Br2N3S.
  • CAS number 142457-00-9 for unambiguous identification.
  • Available in small research quantities, such as 10 MG, for laboratory use.
  • Stable when stored sealed and kept away from moisture; refrigerated storage extends stability.

Catalog No. 50-002-12062 Supplier Medchemexpress LLC Supplier No. HY10106310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amthamine dihydrobromide is the dihydrobromide salt of amthamine, a histamine receptor agonist active at H1-H4 subtypes. It is supplied for research use to investigate histamine receptor pharmacology, gastric acid secretion, and hepatotoxicity models. Follow standard laboratory safety and storage recommendations.

  • High purity (99.6%) suitable for biochemical and pharmacological studies.
  • Molecular weight 319.07 g/mol.
  • Chemical formula C6H13Br2N3S.
  • CAS number 142457-00-9 for unambiguous identification.
  • Available in small research quantities, such as 10 MG, for laboratory use.
  • Stable when stored sealed and kept away from moisture; refrigerated storage extends stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.